Skip to Content
Merck
Methoxyacetaldehyde dimethyl acetal ≥98.0% (GC)

Methoxyacetaldehyde dimethyl acetal

Methoxyacetaldehyde dimethyl acetal ≥98.0% (GC)
Synonym(s):
1,1,2-Trimethoxyethane
Linear Formula:
CH3OCH2CH(OCH3)2
CAS Number:
Molecular Weight:
120.15
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing