
Methotrexate for peak identification

Synonym(s):
Methotrexate, (+)-Amethopterin, 4-Amino-10-methylfolic acid, 4-Amino-N10-methylpteroyl-L-glutamic acid, Antifolan, MTX, Methylaminopterin
Empirical Formula (Hill Notation):
C20H22N8O5
CAS Number:
Molecular Weight:
454.44
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing