
Metacresol

Synonym(s):
m-Cresol, 1-Hydroxy-3-methylbenzene, 3-Hydroxytoluene, 3-Methylphenol
Linear Formula:
CH3C6H4OH
CAS Number:
Molecular Weight:
108.14
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing