
Mesna

Synonym(s):
Sodium 2-mercaptoethanesulfonate, 2-Mercaptoethanesulfonic acid sodium salt, Coenzyme M sodium salt, HS-CoM Na, MESNA
Linear Formula:
HSCH2CH2SO3Na
CAS Number:
Molecular Weight:
164.18
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing