Skip to Content
Merck
Melatonin phyproof® Reference Substance

Melatonin

Melatonin phyproof® Reference Substance
Synonym(s):
N-Acetyl-5-methoxytryptamine, Melatonin
Empirical Formula (Hill Notation):
C13H16N2O2
CAS Number:
Molecular Weight:
232.28

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique