Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnesium D-gluconate hydrate
Synonym(s):
D-Gluconic acid hemimagnesium salt, D-Gluconic acid magnesium salt
Empirical Formula (Hill Notation):
C
12
H
22
MgO
14
· xH
2
O
CAS Number:
3632-91-5
Molecular Weight:
414.60 (anhydrous basis)
EC Number:
222-848-2
Products
Product Category
carbohydrates (2)
disaccharides (1)
monosaccharides (1)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Optical Isomer
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
G9130
Magnesium D-gluconate hydrate,
≥98% (HPLC)
Expand
View Pricing
M7554
Magnesium D-gluconate hydrate,
98.0-102% anhydrous basis, meets USP testing specifications
Expand
View Pricing