
Magnesium citrate tribasic nonahydrate

Synonym(s):
Citric acid magnesium salt, Trimagnesium dicitrate nonahydrate, tri-Magnesium dicitrate nonahydrate
Linear Formula:
Mg3(C6H5O7)2 · 9H2O
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Special Grade
Sterility
Showing 1-1 of 1
Compare
Product Number
Description
Pricing