Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Magnesium carbonate basic
Synonym(s):
Magnesium hydroxide carbonate
Linear Formula:
MgCO
3
CAS Number:
39409-82-0
Molecular Weight:
84.31
EC Number:
235-192-7
Products
Product Category
inorganic bases (3)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Agency
Chemical Composition
Quality Segment
Special Grade
Purum (2)
Purum P.A. (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
63062
Magnesium carbonate basic,
purum p.a., heavy, ≥40% (MgO)
Expand
View Pricing
13118
Magnesium carbonate basic,
purum, light, ≥40% Mg (as MgO) basis, powder (light)
Expand
View Pricing
63032
Magnesium carbonate basic,
tested according to Ph. Eur., heavy
Expand
View Pricing