Lysozyme from chicken egg white
Synonym(s):
Mucopeptide N-acetylmuramoylhydrolase, Muramidase
CAS Number:
EC Number:
Products
Product Category
Brand
Biological Source
Physical Form
Product Line
Purity
Format
Analyte Suitability
Tissue
Chemical Composition
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Sterilization Method
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing