Lorcainide hydrochloride
Synonym(s):
Lorcamide hydrochloride, N-(4-Chlorophenyl)-N-[1-(1-methylethyl)-4-piperidinyl]-benzeneacetamide hydrochloride, R15889 hydrochloride
Empirical Formula (Hill Notation):
C22H27ClN2O · HCl
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1