
L-Menthol

Synonym(s):
(1R,2S,5R)-(−)-Menthol, (−)-Menthol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol, 5-Methyl-2-(1-methylethyl)cyclohexanol, (-)-Menthol, (1R,2S,5R)-2-Isopropyl-5-methylcyclohexanol
Empirical Formula (Hill Notation):
C10H20O
CAS Number:
Molecular Weight:
156.27
Products
Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique