
Lithium metaborate

Synonym(s):
Boric acid lithium salt
Linear Formula:
LiBO2
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing