Skip to Content
Merck
Lithium ionophore VI for ion-selective electrodes, Selectophore™, function tested, ≥99.0% (HPLC)

Lithium ionophore VI

Lithium ionophore VI for ion-selective electrodes, Selectophore™, function tested, ≥99.0% (HPLC)
Synonym(s):
6,6-Dibenzyl-1,4,8-11-tetraoxacyclotetradecane, 6,6-Dibenzyl-14-crown-4
Empirical Formula (Hill Notation):
C24H32O4
CAS Number:
Molecular Weight:
384.51

Products

Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Showing 1-1 of 1