Skip to Content
Merck
Lisinopril Pharmaceutical Secondary Standard; Certified Reference Material

Lisinopril

Lisinopril Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(S)-1-[N2-(1-carboxy-3-phenylpropyl)-lysyl-proline dihydrate, MK-521
Empirical Formula (Hill Notation):
C21H31O5N3 · 2H2O
CAS Number:
Molecular Weight:
441.52

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique