Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lipopolysaccharides from
Escherichia coli
O111:B4
Synonym(s):
LPS
EC Number:
297-473-0
Products
Product Category
carbohydrates (8)
lipopolysaccarides (7)
Brand
Sigma-Aldrich (8)
Biological Source
Color
Physical Form
Product Line
Research Area
Conjugate
Quality Segment
Application
Sterilization Method
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
L2630
Lipopolysaccharides from
Escherichia coli
O111:B4,
purified by phenol extraction
Expand
View Pricing
L4391
Lipopolysaccharides from
Escherichia coli
O111:B4,
γ-irradiated, BioXtra, suitable for cell culture
Expand
View Pricing
L5293
Lipopolysaccharides from
Escherichia coli
O111:B4,
Ready Made solution, 1 mg/mL
Expand
View Pricing
L3024
Lipopolysaccharides from
Escherichia coli
O111:B4,
purified by ion-exchange chromatography, TLR ligand tested
Expand
View Pricing
L3012
Lipopolysaccharides from
Escherichia coli
O111:B4,
purified by gel-filtration chromatography
Expand
View Pricing
L4130
Lipopolysaccharides from
Escherichia coli
O111:B4,
purified by trichloroacetic acid extraction
Expand
View Pricing
F3665
Lipopolysaccharides from
Escherichia coli
O111:B4,
FITC conjugate
Expand
View Pricing
L3023
Lipopolysaccharides from
Escherichia coli
O111:B4,
Detoxified
Expand
View Pricing