
Linezolid

Synonym(s):
N-[[(5S)-3-[3-Fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]acetamide, PNU-100766, U-100766, Linezolid
Empirical Formula (Hill Notation):
C16H20FN3O4
CAS Number:
Molecular Weight:
337.35
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade