Lincomycin hydrochloride
Synonym(s):
Lincomycin hydrochloride monohydrate
Empirical Formula (Hill Notation):
C18H34N2O6S · HCl · H2O
CAS Number:
Molecular Weight:
461.01
Products
Product Category
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3