Skip to Content
Merck
L-Glyceric acid sodium salt ≥95.0% (TLC)

L-Glyceric acid sodium salt

L-Glyceric acid sodium salt ≥95.0% (TLC)
Synonym(s):
(S)-2,3-Dihydroxypropanoic acid sodium salt, Sodium L-glycerate
Empirical Formula (Hill Notation):
C3H6O4 · xNa+
CAS Number:
Molecular Weight:
106.08 (free acid basis)

Products

Product Category
Color
Formula Weight
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing