Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
L-Glutathione reduced
Synonym(s):
γ-L-Glutamyl-L-cysteinyl-glycine, GSH
Linear Formula:
H
2
NCH(CO
2
H)CH
2
CH
2
CONHCH(CH
2
SH)CONHCH
2
CO
2
H
CAS Number:
70-18-8
Molecular Weight:
307.32
EC Number:
200-725-4
Products
Product Category
amino acids (3)
Brand
Sigma-Aldrich (3)
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Optical Isomer
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
G6013
L-Glutathione reduced,
≥98.0%, suitable for cell culture, BioReagent
Expand
View Pricing
G4251
L-Glutathione reduced,
≥98.0%
Expand
View Pricing
G6529
L-Glutathione reduced,
≥98.0%, BioXtra
Expand
View Pricing