
Levofloxacin hemihydrate

Synonym(s):
Levofloxacin hemihydrate
Empirical Formula (Hill Notation):
C18H20FN3O4 · 0.5H2O
CAS Number:
Molecular Weight:
370.38
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2