
Levofloxacin

Synonym(s):
Levofloxacin hemihydrate
Empirical Formula (Hill Notation):
C18H20FN3O4 · 0.5H2O
CAS Number:
Molecular Weight:
370.38
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing