Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Leukemia Inhibitory Factor human
Synonym(s):
LIF, rLIF
Products
Product Category
proteins (3)
growth factors (2)
stem cells (1)
Brand
Sigma-Aldrich (5)
Biological Source
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
LIF1010
Leukemia Inhibitory Factor human,
10 µg, human recombinant LIF protein, expressed in E. coli, suitable for stem cell culture
Expand
View Pricing
LIF1050
Leukemia Inhibitory Factor human,
50 µg, human recombinant LIF protein, expressed in E. coli, suitable for stem cell culture
Expand
View Pricing
L5283
Leukemia Inhibitory Factor human,
≥95% (SDS-PAGE), recombinant, expressed in E. coli, buffered aqueous solution, suitable for cell culture
Expand
View Pricing
LIF1005
Leukemia Inhibitory Factor human,
5 µg, human recombinant LIF protein, expressed in E. coli, suitable for stem cell culture
Expand
View Pricing
SRP9001
Leukemia Inhibitory Factor human,
≥95% (SDS-PAGE), recombinant, expressed in HEK 293 cells, liquid, suitable for cell culture
Expand
View Pricing