Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Leucine Aminopeptidase, microsomal from porcine kidney
Synonym(s):
Aminopeptidase M, Aminopeptidase N, Cathepsin III, LAP, Leucinamide aminopeptidase, Leucinaminopeptidase
CAS Number:
9054-63-1
EC Number:
232-942-5
Products
Product Category
proteases (2)
Brand
Sigma-Aldrich (2)
Biological Source
Physical Form
Tissue
Chemical Composition
Quality Segment
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
L5006
Leucine Aminopeptidase, microsomal from porcine kidney,
Type IV-S, ammonium sulfate suspension, 10-40 units/mg protein (Bradford)
Expand
View Pricing
L6007
Leucine Aminopeptidase, microsomal from porcine kidney,
Type VI-S, lyophilized powder, ≥12 units/mg protein (biuret)
Expand
View Pricing