
Leucine

Synonym(s):
L-Leucine, (S)-2-Amino-4-methylpentanoic acid
Linear Formula:
(CH3)2CHCH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
131.17
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing