
L-(−)-Dithiothreitol

Synonym(s):
(−)-Cleland’s Reagent, (2R,3R)-1,4-Dimercapto-2,3-butanediol, L-DTT, Cleland reagent chiral
Empirical Formula (Hill Notation):
C4H10O2S2
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing