
L-Buthionine-sulfoximine

Synonym(s):
L-BSO
Empirical Formula (Hill Notation):
C8H18N2O3S
CAS Number:
Molecular Weight:
222.31
Products
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing