
Lasalocid A sodium salt

Synonym(s):
Lasalocid sodium salt
Empirical Formula (Hill Notation):
C34H53NaO8
CAS Number:
Molecular Weight:
612.77
Products
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Activity Spectrum
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing