
Lapatinib

Synonym(s):
Lapatinib
Empirical Formula (Hill Notation):
C29H26ClFN4O4S
CAS Number:
Molecular Weight:
581.06
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Showing 1-2 of 2
Compare
Product Number
Description
Pricing