Skip to Content
Merck
vibrant-m

Lansoprazole Related Compound B

vibrant-m
Synonym(s):
2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)-pyridin-2-yl]methyl]sulfanyl]-1H-benzimidazole, Lansoprazole sulfide
Empirical Formula (Hill Notation):
C16H14F3N3OS
CAS Number:
Molecular Weight:
353.36

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1