
L-α-Hydroxyglutaric acid disodium salt

Synonym(s):
(S)-2-Hydroxypentanedioic acid disodium salt, Disodium (S)-2-hydroxyglutarate
Linear Formula:
C5H6O5Na2
CAS Number:
Molecular Weight:
192.08
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Optical Isomer
Relevant Disease
Showing 1-1 of 1
Compare
Product Number
Description
Pricing