Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Lactococcus lactis
subsp.
lactis
WDCM 00016 Vitroids
™
Products
Brand
Millipore (2)
Manufacturer
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Special Grade
certified reference material (2)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
VT000167
Lactococcus lactis
subsp.
lactis
WDCM 00016 Vitroids
™
,
50’000-150’000 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing
VT000163
Lactococcus lactis
subsp.
lactis
WDCM 00016 Vitroids
™
,
80-130 CFU mean value range, certified reference material, suitable for microbiology, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing