Skip to Content
Merck
Lactic acid solution 10%, suitable for microbiology, sterile filtered

Lactic acid solution

Lactic acid solution 10%, suitable for microbiology, sterile filtered
Linear Formula:
CH3CH(OH)CO2H
CAS Number:
Molecular Weight:
90.08

Products

Product Category
Brand
Formula Weight
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2