
Lacosamide-13C, D3 solution

Empirical Formula (Hill Notation):
13CC12D3H15N2O3
Molecular Weight:
254.30
EC Number:
Products
Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique