Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
K-252a
Empirical Formula (Hill Notation):
C
27
H
21
N
3
O
5
CAS Number:
99533-80-9
Molecular Weight:
467.47
Products
Product Category
bioactive small molecules (2)
Brand
Sigma-Aldrich (2)
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
K2015
K-252a,
Ready Made Solution, from
Nonomuraea longicatena
, >98%
Expand
View Pricing
K1639
K-252a,
from
Nonomuraea longicatena
, ≥98% (HPLC)
Expand
View Pricing