
Jones reagent

Synonym(s):
Jone′s reagent, Chromium trioxide solution in sulfuric acid, Sulfuric acid-chromium trioxide mixture
CAS Number:
Products
Brand
Physical Form
Chemical Composition
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing