
Isopropylmagnesium chloride solution

Linear Formula:
(CH3)2CHMgCl
CAS Number:
Molecular Weight:
102.85
Products
Product Category
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing