
Isopropyl chloroformate solution

Linear Formula:
(CH3)2CHOCOCl
CAS Number:
Molecular Weight:
122.55
Products
Product Category
Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing