Skip to Content
Merck
Isobutyl propionate ≥98%, FG

Isobutyl propionate

Isobutyl propionate ≥98%, FG
Linear Formula:
CH3CH2CO2CH2CH(CH3)2
CAS Number:
Molecular Weight:
130.18
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing