Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Iron/Nickel/Boron
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GF88131832
Iron/Nickel/Boron,
foil, thickness 0.025 mm, Fe40%/Ni38%/B 18%/Mo 4%, length 0.5 m, coil width 50 mm
Expand
View Pricing
GF59913928
Iron/Nickel/Boron,
foil, Fe40%/Ni38%/B 18%/Mo 4%, thickness 0.025 mm, coil width 50 mm, length 2 m
Expand
View Pricing
GF16479311
Iron/Nickel/Boron,
foil, Fe 40%/Ni 38%/B 18%/Mo 4%, thickness 0.025 mm, length 1 m, coil width 50 mm
Expand
View Pricing
GF55095857
Iron/Nickel/Boron,
foil, Fe 40%/Ni 38%/B 18%/Mo 4%, thickness 0.025 mm, length 10 m, coil width 50 mm
Expand
View Pricing
GF89240766
Iron/Nickel/Boron,
foil, thickness 0.025 mm, Fe40%/Ni38%/B 18%/Mo 4%, length 5 m, coil width 50 mm
Expand
View Pricing