Skip to Content
Merck
Iridium wire reel, 2m, diameter 0.15mm, as drawn, 99.9%

Iridium

Iridium wire reel, 2m, diameter 0.15mm, as drawn, 99.9%
Synonym(s):
Iridium
Empirical Formula (Hill Notation):
Ir
CAS Number:
Molecular Weight:
192.22

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Diameter (mm)
Length (m)
Element
Showing 1-20 of 68
Page 1 of 4
Compare
Product Number
Description
Pricing
Page 1 of 4