
Iohexol

Synonym(s):
5-[N-(2,3-Dihydroxypropyl)acetamido]-2,4,6-triiodo-N,N′-bis(2,3-dihydroxypropyl)isophthalamide
Empirical Formula (Hill Notation):
C19H26I3N3O9
CAS Number:
Molecular Weight:
821.14
Products
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Technique
Showing 1-3 of 3