Skip to Content
Merck
Interleukin-12 human ≥97% (SDS-PAGE and N-terminal analysis), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture

Interleukin-12 human

Interleukin-12 human ≥97% (SDS-PAGE and N-terminal analysis), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture
Synonym(s):
IL-12

Products

Product Category
Brand
Biological Source
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Showing 1-1 of 1