Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Indium-tin alloy, In90Sn10
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Length (mm)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
GF71962487
Indium-tin alloy, In90Sn10,
50mm rod, 5.0mm diameter
Expand
View Pricing
GF68821937
Indium-tin alloy, In90Sn10,
200mm rod, 5.0mm diameter
Expand
View Pricing
GF39051985
Indium-tin alloy, In90Sn10,
sheet, 50 x 50mm, 3.0mm thickness
Expand
View Pricing
GF61873241
Indium-tin alloy, In90Sn10,
100mm rod, 5.0mm diameter
Expand
View Pricing
GF40351441
Indium-tin alloy, In90Sn10,
sheet, 50 x 50mm, 2.0mm thickness
Expand
View Pricing
GF65667842
Indium-tin alloy, In90Sn10,
sheet, 100 x 100mm, 2.0mm thickness
Expand
View Pricing