
Indium(III) phthalocyanine chloride

Empirical Formula (Hill Notation):
C32H16ClInN8
CAS Number:
Molecular Weight:
662.79
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Showing 1-2 of 2
Compare
Product Number
Description
Pricing