
IL-4 human

Synonym(s):
B cell stimulating factor (BSF-1), BCDF, BCGF, B cell stimulating factor, BSF-1, IL-4, Lymphocyte stimulatory factor 1
Products
Product Category
Brand
Biological Source
Color
Gene Alias
Physical Form
Purity
Recombinant Host
Quality Segment
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing