
Ibuprofen

Synonym(s):
α-Methyl-4-(isobutyl)phenylacetic acid, (±)-2-(4-Isobutylphenyl)propanoic acid, (±)-Ibuprofen
Empirical Formula (Hill Notation):
C13H18O2
Products
Product Category
Brand
Color
Feature
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing