
Hypoxanthine

Synonym(s):
6-Hydroxypurine, Hypoxanthine
Empirical Formula (Hill Notation):
C5H4N4O
CAS Number:
Molecular Weight:
136.11
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing