
Hygromycin B

Synonym(s):
Hygromycin B from Streptomyces hygroscopicus, Hygromycin B, Streptomyces sp.
Empirical Formula (Hill Notation):
C20H37N3O13
CAS Number:
Molecular Weight:
527.52
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Chemical Composition
Quality Segment
Mode of Action
Sterility
Technique