Skip to Content
Merck
Hydroquinone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Hydroquinone

Hydroquinone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
Hydroquinone, 1,4-Benzenediol, 1,4-Dihydroxybenzene, HQ
Linear Formula:
C6H4-1,4-(OH)2
CAS Number:
Molecular Weight:
110.11
EC Number:

Products

Brand
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
Package Size
Reagent Type
API Family
Reference Material Type
Special Grade
Technique