Skip to Content
Merck
Hydromorphone solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Hydromorphone solution

Hydromorphone solution 1 mg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®
Synonym(s):
Hydromorphone solution
Empirical Formula (Hill Notation):
C17H19NO3
CAS Number:
Molecular Weight:
285.34

Products

Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade